Anti-MARK1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7144BEA 348 EUR
ABGEAP7144B
Anti-MARK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARK1 (MAP/microtubule affinity-regulating kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR