Anti-STRADA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7142BEA 350 EUR
ABGEAP7142B
Anti-STRADA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STRADA (STE20-related kinase adaptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR