Anti-Guanylyl Cyclase alpha 2 (GUCY1A2) Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7133AEA 350 EUR
ABGEAP7133A
Anti-Guanylyl Cyclase alpha 2 (GUCY1A2) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Guanylyl Cyclase alpha 2 (GUCY1A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR