Anti-Ketohexokinase (KHK) Rabbit Polyclonal Antibody
Lieferant: Abgent
ABGEAP7069AEA 350 EUR
ABGEAP7069A
Anti-Ketohexokinase (KHK) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Ketohexokinase (KHK)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse