Anti-PDK4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7041BEA 348 EUR
ABGEAP7041B
Anti-PDK4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDK4 (pyruvate dehydrogenase kinase, isozyme 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR