Anti-PKC beta1/2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7016AEA 350 EUR
ABGEAP7016A
Anti-PKC beta1/2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PKC beta1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR