Anti-RPS6KA4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7011AEA 360 EUR
ABGEAP7011A
Anti-RPS6KA4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS6KA4 (ribosomal protein S6 kinase, 90kDa, polypeptide 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR