Anti-ADRBK1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7004AEA 348 EUR
ABGEAP7004A
Anti-ADRBK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADRBK1 (adrenergic, beta, receptor kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR