Anti-AURKA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7002CEA 360 EUR
ABGEAP7002C
Anti-AURKA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AURKA (aurora kinase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR