Anti-NAT2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6993BEA 348 EUR
ABGEAP6993B
Anti-NAT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAT2 (N-acetyltransferase 2 (arylamine N-acetyltransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR