Anti-ITGAX Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6986BEA 360 EUR
ABGEAP6986B
Anti-ITGAX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR