Anti-RAB5A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6974BEA 360 EUR
ABGEAP6974B
Anti-RAB5A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAB5A (RAB5A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR