Anti-MT-ND5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6939BEA 350 EUR
ABGEAP6939B
Anti-MT-ND5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MT-ND5 (mitochondrially encoded NADH dehydrogenase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR