Anti-HADHA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6882BEA 0 EUR
ABGEAP6882B
Anti-HADHA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HADHA (hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR