Anti-SOD3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6871AEA 348 EUR
ABGEAP6871A
Anti-SOD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOD3 (superoxide dismutase 3, extracellular)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR