Anti-IL1A-S87 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6860AEA 350 EUR
ABGEAP6860A
Anti-IL1A-S87 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL1A-S87
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR