Anti-FHL1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6852BEA 350 EUR
ABGEAP6852B
Anti-FHL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FHL1 (four and a half LIM domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR