Anti-FGA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6847AEA 360 EUR
ABGEAP6847A
Anti-FGA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGA (fibrinogen alpha chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR