Anti-CDK14 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6780AEA 350 EUR
ABGEAP6780A
Anti-CDK14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK14 (cyclin-dependent kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR