Anti-FBP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6729BEA 350 EUR
ABGEAP6729B
Anti-FBP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FBP2 (fructose-1,6-bisphosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR