Anti-LRG1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6705AEA 348 EUR
ABGEAP6705A
Anti-LRG1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRG1 (leucine-rich alpha-2-glycoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR