Anti-PSME2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6692BEA 348 EUR
ABGEAP6692B
Anti-PSME2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSME2 (proteasome (prosome, macropain) activator subunit 2 (PA28 beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR