Anti-XPO1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6684BEA 348 EUR
ABGEAP6684B
Anti-XPO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XPO1 (exportin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR