Anti-TBP Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6680BEA 360 EUR
ABGEAP6680B
Anti-TBP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TBP (TATA box binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR