Anti-PRELP Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6665BEA 350 EUR
ABGEAP6665B
Anti-PRELP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRELP (proline/arginine-rich end leucine-rich repeat protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR