Anti-HMGCS1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6652BEA 350 EUR
ABGEAP6652B
Anti-HMGCS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HMGCS1 (3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR