Anti-GLS2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6650DEA 360 EUR
ABGEAP6650D
Anti-GLS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GLS2 (glutaminase 2 (liver, mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR