Anti-RPS9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6618BEA 350 EUR
ABGEAP6618B
Anti-RPS9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS9 (ribosomal protein S9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR