Anti-BAALC Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6605AEA 348 EUR
ABGEAP6605A
Anti-BAALC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BAALC (brain and acute leukemia, cytoplasmic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR