Anti-WNT10B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6595BEA 348 EUR
ABGEAP6595B
Anti-WNT10B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WNT10B (wingless-type MMTV integration site family, member 10B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR