Anti-GZMB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6575BEA 350 EUR
ABGEAP6575B
Anti-GZMB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GZMB (granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR