Anti-IGJ Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6548AEA 350 EUR
ABGEAP6548A
Anti-IGJ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGJ (immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR