Anti-DNAJC19 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6542CEA 350 EUR
ABGEAP6542C
Anti-DNAJC19 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNAJC19 (DnaJ (Hsp40) homolog, subfamily C, member 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR