Anti-CLNS1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6520BEA 350 EUR
ABGEAP6520B
Anti-CLNS1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLNS1A (chloride channel, nucleotide-sensitive, 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR