Anti-SNCA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6404AEA 350 EUR
ABGEAP6404A
Anti-SNCA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNCA (synuclein, alpha (non A4 component of amyloid precursor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR