Anti-Myosin 1C (MYO1C) Rabbit Polyclonal Antibody
Lieferant: Abgent
ABGEAP6372BEA 350 EUR
ABGEAP6372B
Anti-Myosin 1C (MYO1C) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Myosin 1C (MYO1C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human