Anti-TAP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6252AEA 350 EUR
ABGEAP6252A
Anti-TAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAP1 (transporter 1, ATP-binding cassette, sub-family B (MDR/TAP))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR