Anti-MMP10 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6194AEA 350 EUR
ABGEAP6194A
Anti-MMP10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP10 (matrix metallopeptidase 10 (stromelysin 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR