Anti-MLLT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6188AEA 360 EUR
ABGEAP6188A
Anti-MLLT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MLLT1 (myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR