Anti-MAGEL2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6181AEA 348 EUR
ABGEAP6181A
Anti-MAGEL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAGEL2 (MAGE-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR