Anti-LPAR2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6140AEA 360 EUR
ABGEAP6140A
Anti-LPAR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LPAR2 (lysophosphatidic acid receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR