Anti-GREM1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6133AEA 360 EUR
ABGEAP6133A
Anti-GREM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GREM1 (gremlin 1, DAN family BMP antagonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR