Anti-ABCB5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP6122AEA 348 EUR
ABGEAP6122A
Anti-ABCB5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABCB5 (ATP-binding cassette, sub-family B (MDR/TAP), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR