Anti-HTR3C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5973CEA 350 EUR
ABGEAP5973C
Anti-HTR3C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTR3C (5-hydroxytryptamine (serotonin) receptor 3C, ionotropic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR