Anti-ATP5D Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5967AEA 348 EUR
ABGEAP5967A
Anti-ATP5D Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATP5D (ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR