Anti-PTGR1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5941CEA 348 EUR
ABGEAP5941C
Anti-PTGR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTGR1 (prostaglandin reductase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR