Anti-SLC16A9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5934CEA 350 EUR
ABGEAP5934C
Anti-SLC16A9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC16A9 (solute carrier family 16, member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR