Anti-SMS Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5883AEA 350 EUR
ABGEAP5883A
Anti-SMS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMS (spermine synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR