Anti-LRRC52 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5857BEA 348 EUR
ABGEAP5857B
Anti-LRRC52 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRRC52 (leucine rich repeat containing 52)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR