Anti-GCH1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP5839AEA 350 EUR
ABGEAP5839A
Anti-GCH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GCH1 (GTP cyclohydrolase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR